Learn More
Medchemexpress LLC CaMKIIα-IN-1 | 3031484-08-6 | 98.4% | 278.69 g/mol | C14H11ClO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1462685MG
Encompass_Preferred
CaMKIIα-IN-1 is an orally active inhibitor of Ca2+/calmodulin-dependent protein kinase II alpha (CaMKIIα). It binds the CaMKIIα WT hub with a reported KD of 219 nM and shows good metabolic stability. The compound is provided at high purity for research applications and is available in small-scale quantities suitable for biochemical and cellular studies.
  • High binding affinity to CaMKIIα (KD ≈ 219 nM).
  • Orally active in preclinical studies.
  • Demonstrated good metabolic stability.
  • High purity suitable for research (≈98.4%).
  • Available in small-scale quantities for assay development.
Catalog No. 50-002-23086