Learn More
Selleck Chemical LLC Camostat Mesilate S2874-10mg
SDP
Supplier:  Selleck Chemical LLC S287410MG
Encompass_Preferred
Camostat (FOY-305 FOY-S980) is a trypsin-like protease inhibitor inhibits airway epithelial sodium channel (ENaC) function with IC50 of 50 nM less potent to trpsin prostasin and matriptase
Catalog No. 50-311-1865