Learn More
Apexbio Technology LLC Camostat Mesilate 59721-29-8 10mg
SDP
Supplier:  Apexbio Technology LLC B208210
Encompass_Preferred
Camostat Mesilate (59721-29-8) is a small-molecule inhibitor targeting trypsin-like serine proteases It is designed to suppress protease activity thereby modulating processes such as epithelial sodium channel (ENaC) function and airway surface liquid homeostasis Camostat Mesilate exerts its biological activity primarily through protease inhibition In cell-based studies using airway epithelial cells Camostat Mesilate demonstrates ENaC activity suppression with an IC50 value of 50 nM Based on these pharmacological properties Camostat Mesilate holds research potential in studies of respiratory physiology and pathophysiology particularly those investigating ENaC-mediated ion transport in airway epithelial cell models
Catalog No. 50-000-90079