Learn More
Medchemexpress LLC Camostat mesylate | 59721-29-8 | MFCD00941410 | >98.0% | 494.52 | C21H26N4O8S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY135125MG
Encompass_Preferred
Camostat mesylate is an orally active, synthetic serine protease inhibitor supplied as the mesylate salt. It inhibits proteases such as trypsin, prostasin, matriptase, and TMPRSS2, and is used in enzymatic and cell-based studies of protease function and viral entry. CAS: 59721-29-8; molecular weight: 494.52.
  • Orally active serine protease inhibitor used in research.
  • Inhibits TMPRSS2, useful for viral entry and antiviral studies.
  • Suitable for enzyme inhibition and cell-based assays.
  • Provided as a mesylate salt for enhanced stability and handling.
  • Available in small research quantities for assay development.
Catalog No. 50-002-23099