Learn More
Abcam Camostat mesylate, TMPRSS2 inhibitor, 10MG
Supplier:  Abcam AB14570910MG
Encompass_Preferred
MW 494.52 g/mol, Purity >98%. Trypsin-like protease inhibitor. Reversibly inhibits airway epithelial sodium channel (ENaC) function (IC₅₀ = 50 nM). Inhibits the production of TNF-α and monocyte chemoattractant protein-1 (MCP-1) by monocytes and disrupts proliferation of pancreatic stellate cells in a rat model of pancreatic fibrosis. Inhibits transmembrane serine protease TMPRSS2 and blocks entry of SARS-Cov-2 into lung cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5080