Learn More
Enzo Life Sciences Enzo Life Sciences Direct cAMP ELISA Kit, Sensitive Assay for Cyclic AMP, Colorimetric Detection, Kit
SDP
Supplier:  Enzo Life Sciences ADI900067A
Encompass_Preferred
The Enzo Life Sciences Direct cAMP ELISA Kit offers a sensitive and reliable method for quantitatively measuring cyclic AMP (cAMP) levels in various biological samples. With its unique acetylation reagents, it enhances sensitivity dramatically, making it ideal for low-level analyses. This competitive colorimetric immunoassay simplifies sample preparation by allowing direct analysis, minimizing errors and ensuring consistent results, setting it apart from traditional methods.
Catalog No. NC2068020