Learn More
Enzo Life Sciences Adenosine 3',5'-cyclic Monophosphate. sodium salt (100mg). CAS: 37839-81-9
SDP
Supplier:  Enzo Life Sciences ALX480011M100
Encompass_Preferred
Key regulator of many cellular reactions. Naturally occurring activator of cAMP-dependent protein kinase (PKA). Alternative name: Cyclic AMP . Na, cAMP . Na. Purity: ≥98% (HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9303