Learn More
MSC Campbell Clevis Hook, 1/2 in Chain Diameter, Grade 80, Alloy Steel
Supplier:  MSC 67782565
Encompass
This clevis hook is designed for heavy-duty lifting applications, featuring a 1/2 in chain diameter and a Grade 80 rating for superior strength and durability. Constructed from heat-treated painted alloy steel, it offers a high work load limit of 13,000 lb, making it suitable for demanding industrial environments. The hook's design includes a 9/16 in clevis pin diameter and a 1.75 in hook throat, ensuring secure connections and reliable performance.
  • Grade 80 chain for high strength applications.
  • 13,000 lb work load limit.
  • Constructed from heat-treated painted alloy steel for durability.
  • 9/16 in clevis pin diameter.
  • 1.75 in hook throat.
  • Designed for 1/2 in chain diameter.
Catalog No. 50-005-31051