Learn More
MSC Campbell Clevis Hook, 3/4 in Chain Dia, Grade 70, Alloy Steel
Supplier: MSC 67782086
This clevis hook is designed for heavy-duty applications, featuring a 3/4 inch chain diameter and constructed from durable alloy steel. It is rated for Grade 70 and offers a substantial work load limit.
- Grade 70 for high strength applications.
- Work load limit of 28,300 lb.
- Constructed from alloy steel for durability.
- Designed for 3/4 inch chain diameter.
- Features a 15/16 inch clevis pin diameter.
- Hook throat opening is 0.9375 "