Learn More
MSC Campbell S-Hook, Carbon Steel, Zinc Plated, 0.175 in Wire, 95 lb Capacity
Supplier:  MSC 67788182
Encompass
The Campbell S-Hook is a robust connecting component made from carbon steel with a zinc-plated finish. Designed for various applications, it features a 0.175 in wire diameter and an overall length of 1-3/8 in. This S-Hook provides a reliable connection point with a load capacity of 95 lb.
  • Carbon steel construction for durability.
  • Zinc plated finish for corrosion resistance.
  • Wire size of 0.175 in accommodates various connections.
  • Overall length of 1-3/8 in for versatile use.
  • Safely handles loads up to 95 lb.
Catalog No. 50-005-30916