Learn More
Selleck Chemical LLC ()-Camphor S4516-100mg
SDP
Supplier:  Selleck Chemical LLC S4516100MG
Encompass_Preferred
Camphor is a waxy flammable white or transparent solid with a strong aroma often used to relieve pain reduce itching and treat fungal infections It can activate TRPV1 and TRPV3
Catalog No. 50-311-0407