Learn More
Medchemexpress LLC Camphor (Standard) | 76-22-2 | 152.23 | 50 MG

Supplier: Medchemexpress LLC HYN0808R50MG
Camphor (Standard) is the analytical standard of Camphor, intended for research and analytical applications. Camphor ((±)-Camphor) is a topical anti-infective and anti-pruritic, and internally acts as a stimulant and carminative. It is poisonous when ingested and exhibits antiviral, antitussive, and anticancer activities. Camphor is also a TRPV3 agonist.
- Purity: 99.86%
- Application: The compound is an analytical standard, a reference standard supplied assay. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
- Appearance: Solid
- Color: White to off-white
- Initial source: Plants Lauraceae Cinnamomum camphora
- Shipping: Room temperature in continental US; may vary elsewhere.
- Storage: 4°C, sealed storage, away from light and moisture.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.