Learn More
Selleck Chemical LLC Camptothecin 100mg 7689-03-4 NSC-100880, CPT
SDP
Supplier:  Selleck Chemical LLC S1288100MG
Encompass_Preferred
Camptothecin (NSC-100880, CPT) is a specific inhibitor of DNA topoisomerase I (Topo I) with IC50 of 0.68 ?M in a cell-free assay. Camptothecin induces apoptosis in cancer cells via microRNA-125b-mediated mitochondrial pathways. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1118