Learn More
Medchemexpress LLC HY-16560 100mg , Camptothecin CAS:7689-03-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY16560100MG
Encompass_Preferred
HY-16560 100mg Camptothecin CAS: 7689-03-4 Camptothecin (CPT), a kind of alkaloid, is a DNA topoisomerase I (Topo I) inhibitor with an IC50 of 679 nM. Camptothecin (CPT) exhibits powerful antineoplastic activity against colorectal, breast, lung and ovarian cancers, modulates hypoxia-inducible factor-1α (HIF-1α) activity by changing microRNAs (miRNA) expression patterns in human cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9014