Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Camptothecin-20S-O-p 100mg | 194414-69-2 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY114668100MG

Encompass_Preferred

Camptothecin-20(S)-O-propionate (CZ48) is the C20-propionate ester prodrug of camptothecin, acting as a topoisomerase I inhibitor with reported anticancer activity. It is supplied as a solid research compound for use in in vitro and in vivo studies.

  • High purity: 99.6%.
  • Molecular formula C23H20N2O5 and molecular weight 404.42 g/mol.
  • Used as a topoisomerase I inhibitor in anticancer research.
  • Supplied as a solid suitable for in vitro and in vivo applications.
  • Storage: powder at -20°C for long-term; in solution store at -80°C for short-term.

Catalog No. 50-002-28834


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.