Learn More
Medchemexpress LLC Camptothecin-20S-O-p 10mg | 194414-69-2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11466810MG
Encompass_Preferred
Camptothecin-20(S)-O-propionate (CZ48) is the C20-propionate ester of camptothecin and acts as a topoisomerase I inhibitor used in anticancer research. It is provided as a solid and in ready-to-use DMSO solutions for laboratory studies; intended for research use only.
  • Topoisomerase I inhibitor with a C20-propionate modification.
  • Supplied as solid and 10 mM solutions in DMSO for convenience.
  • High purity suitable for analytical and biological studies.
  • Molecular weight 404.42 g/mol and formula C23H20N2O5.
  • Intended for research use only.
Catalog No. 50-002-29700