Learn More
Medchemexpress LLC Camptothecin-20S-O-p 5mg | 194414-69-2 | 404.42 g/mol | C23H20N2O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1146685MG
Encompass_Preferred
Camptothecin-20(S)-O-propionate (CZ48) is the C20-propionate ester prodrug of camptothecin. It is described as a potent anticancer agent that acts by stabilizing topoisomerase I-DNA covalent complexes, impairing DNA religation and leading to cell death. Supplied for laboratory research in solid and solution forms for in vitro and in vivo studies.
  • Prodrug of camptothecin with the C20 propionate modification.
  • Topoisomerase I inhibitor mechanism of action.
  • High reported purity suitable for research applications.
  • Available as small solid quantities and as a DMSO solution for dosing or assay preparation.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-29477