Learn More
ATCC Genomic DNA from Campylobacter jejuni subsp. jejuni (NCTC 11168), 5 μg, dried
Supplier:  ATCC 700819D5
Encompass
Genomic DNA isolated from Campylobacter jejuni subsp. jejuni (strain NCTC 11168). Supplied as a high-molecular-weight dried genomic DNA preparation (approximately 5 μg total DNA) for PCR, sequencing, and other molecular biology research applications. Store refrigerated and rehydrate before use following the provided instructions. This product is for research use only and is not intended for diagnostic or therapeutic purposes.
  • High molecular weight genomic DNA suitable for PCR and sequencing.
  • Approximately 5 μg total DNA measured by PicoGreen.
  • Supplied dried to facilitate storage and transport.
  • Rehydrate with molecular-grade water; incubate per instructions to improve recovery.
  • Store refrigerated at 2°C to 8°C until rehydration.
  • For research use only; not for human or animal therapeutic or diagnostic use.
Catalog No. NC3238052