Learn More
ATCC Campylobacter upsaliensis; genomic DNA
Supplier:  ATCC BAA1059D5
Encompass
Genomic DNA isolated from Campylobacter upsaliensis Strain RM3195. This bacterial strain is also available as ATCC Catalog No. BAA-1059™.
Catalog No. 50-238-1576