Learn More
Medchemexpress LLC Can f 4 Canine HEK 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7731050UG
Encompass_Preferred
Can f 4 Protein Canine (HEK293 His) an IgE-binding dog dander protein is an important respiratory allergen Can f 4 Protein is a dog allergen of the lipocalin family[1] Can f 4 Protein Canine (HEK293 His) is the recombinant canine-derived Can f 4 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-87235