Learn More
Cosmo Bio Usa Inc CAN GET SIGNAL 250MLCAN GET SI
SDP
Supplier:  Cosmo Bio Usa Inc TYBNKB301
Encompass
Accelerator solution for antigen-antibody reactions. Improves sensitivity, specificity, and S/N for antibody blotting, ELISA, etc. For tissue immunostaining use Can Get Signal Immunostain products.
Catalog No. NC0123544