Learn More
Medchemexpress LLC HY-10451 100mg , Canagliflozin JNJ 24831754ZAE;JNJ 28431754;JNJ 28431754AAA;TA 7284 CAS:842133-18-0 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY10451100MG
Encompass_Preferred
Medchemexpress, HY-10451 100mg Canagliflozin JNJ 24831754ZAE;JNJ 28431754;JNJ 28431754AAA;TA 7284 CAS:842133-18-0 Formula:C24H25FO5S IC50: 2/3.7/4.4 nM (m/r/hSGLT2, in CHOK cells) Purity:98% Canagliflozin is a selective SGLT2 inhibitor with IC 50 of 2 nM, 3.7 nM, and 4.4 nM for mSGLT2, rSGLT2, and hSGLT2 in CHOK cells, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2709