Learn More
Medchemexpress LLC Canagliflozin Standa 25mg | 842133-18-0 | 25MG
SDP
Supplier:  Medchemexpress LLC HY10451R25MG
Encompass_Preferred
Canagliflozin (Standard) is the analytical standard of Canagliflozin This product is intended for research and analytical applications Canagliflozin (JNJ 28431754) is a selective SGLT2 inhibitor with IC50s of 2 nM 3 7 nM and 4 4 nM for mSGLT2 rSGLT2 and hSGLT2 in CHOK cells respectively[1
Catalog No. 50-002-42007