Learn More
Apexbio Technology LLC Candesartan Cilexetil 145040-37-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B22035.1
Encompass_Preferred
Candesartan Cilexetil (CAS 145040-37-5) is a selective angiotensin II receptor antagonist that inhibits the binding of angiotensin II to the AT1 receptor subtype By blocking this interaction it modulates pathways involved in vascular tone and blood pressure regulation In preclinical research Candesartan Cilexetil has been investigated for its capacity to attenuate the progression of pulmonary fibrosis modulate host response in viral infections and facilitate skin wound healing These properties support its utility in studies of cardiovascular pathophysiology fibrotic disease mechanisms viral infection dynamics and tissue repair processes
Catalog No. 50-000-89189