Learn More
ATCC Quantitative Genomic DNA from; Candida parapsilosis; Strain CBS 604
Supplier:  ATCC 22019DQ
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load.
Catalog No. 50-237-9613