Learn More
Medchemexpress LLC Canertinib | 267243-28-7 | MFCD09837878 | 99.0% | 485.94 g/mol | C24H25ClFN5O3 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY10367200MG
Encompass_Preferred
Canertinib is an irreversible epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor used as a research compound to probe EGFR/ErbB signaling. It inhibits cellular EGFR and ErbB2 autophosphorylation at low nanomolar concentrations and is provided as a solid for laboratory use.
  • Irreversible EGFR/ErbB inhibitor with reported IC50s ~7.4 nM (EGFR) and ~9 nM (ErbB2).
  • Suitable for in vitro and in vivo research on EGFR-driven signaling pathways.
  • Provided as a solid, soluble in DMSO for preparation of stock solutions.
  • Molecular formula C24H25ClFN5O3 and molecular weight ~485.94 g/mol.
  • Intended for research use only; not for human or clinical applications.
Catalog No. 50-002-46903