Learn More
Abcam Canine Cardiac Troponin I ELISA Kit - Intracellular, 1 x 96 Tests
Supplier:  Abcam AB3093071X96TESTS
Encompass_Preferred
Canine Cardiac Troponin I ELISA Kit - Intracellular is a Sandwich (quantitative) ELISA kit for the measurement of Canine Cardiac Troponin I - Intracellular in Dog, Monkey, Human in Tissue Extracts samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-9897