Learn More
Raybiotech Inc G-Series Canine Cytokine Array 3 Kit.
Supplier:  Raybiotech Inc GSCCYT31
Encompass_Preferred
Detects 10 Canine Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: ErbB3 (Her3), Galectin-3, GASP-1, IGFBP-2, KC (CXCL1), MMP-8, NCAM-1 (CD56), Nope, PDGF-BB, TGF-alpha
Catalog No. 50-195-0342