Learn More
ACROBiosystems ACROBiosystems Canine IL-13 Protein, His Tag
Supplier:  ACROBiosystems IL3C52H450UG
Encompass_Preferred
Recombinant Protein;50UG;Canine IL-13, His Tag (IL3-C52H4) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Arg 131 (Accession # NP_001003384.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.3 kDa.
Catalog No. 50-210-6393