Learn More
American Research Products Inc Canine Luteinizing Hormone LH
Supplier:  American Research Products Inc CSBE06790C
Encompass
Canine Luteinizing Hormone LH ELISA Kit from CUSABIO Detection Range 0 75 mIU/mL-60 mIU/mL Sensitivity 0 94 mIU/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29928