Learn More
American Research Products Inc Canine N terminal pro brain na
Supplier:  American Research Products Inc CSBE17577C
Encompass
Canine N-terminal pro-brain natriuretic peptide (NT-proBNP) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-27035