Learn More
MOLECULAR DEPOT LLC Charcoal Dextran Serum - 25 mL
Supplier:  MOLECULAR DEPOT LLC B2013910
Encompass_Preferred
Pooled Human Charcoal Dextran Stripped Serum is a human pooled serum that has been treated with Charcoal Dextran to remove lipophilic molecules such as hormones and steroids This product has been used as molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only
Catalog No. 50-000-13487