Learn More
Cell Biologics CANINE PRIMRY PROXIMAL TUB EPI
SDP
Supplier:  Cell Biologics D6015
Encompass_Preferred
Canine Primary Proximal Tubular Epitelia Cells from are isolated from the proximal tubular tissue of beagle dog and grown in gelatin pre-coated tissue culture flask with Complete Growth Medium.
Catalog No. NC2402322