Learn More
American Research Products Inc Canine PTH ELISA Kit
Supplier: American Research Products Inc RDRPTHC
This Canine Parathyroid Hormone (PTH) ELISA Kit is a precoated Competitive Inhibition ELISA assay designed to detect and quantify the target analyte in serum plasma and other biological fluids This product is for Research Use Only