Learn More
Equitech Bio Inc Canine Serum Albumin, High-Purity Protein, Not for In Vivo Use, 500 mg
SDP
Supplier:  Equitech Bio Inc CASA62
Encompass_Preferred
Equitech-Bio Inc. Canine Serum Albumin is a high-purity lyophilized powder intended for various biochemical applications in scientific research. With a purity of ≥ 96% and an undetectable IgG level, it ensures reliable performance and is specifically meant for in vitro use. Its unique preparation through heat-shock fractionation distinguishes it from other similar products, offering researchers a trusted option for canine serum studies.
Catalog No. NC2391504