Learn More
Medchemexpress LLC Cannflavin A | 76735-57-4 | ≥98.0% | 436.50 g·mol⁻¹ | C26H28O6 | 1mg
SDP
Supplier:  Medchemexpress LLC HYW7485911MG
Encompass_Preferred
Cannflavin A can be isolated from Cannabis sativa L Cannflavin A has anti-cancer neuroprotective and anti-inflammatory activity Cannflavin A inhibits A 1-42 aggregation Cannflavin A also inhibits kynurenine-3-monooxygenase (KMO) (IC50 29 4 M) Cannflavin A activates apoptosis via caspase-3 cleavage Cannflavin A exerts anti-inflammatory effects by inhibiting pro-inflammatory enzymes including prostaglandin E2 and cytochrome c oxidases I and II in PC12 cell line[1][2][3][4]
Catalog No. 50-002-70419