Learn More
Sklar Instruments Cannula Instrument Cleaning Brush, 2mm x 12 in., Pack of 3.
SDP
Supplier:  Sklar Instruments 101350
Encompass_Preferred
Cannula Instrument Cleaning Brush, 2mm x 12 in., Pack of 3. Sklar's Cannula Instrument Cleaning Brushes are used in the multiple-step process of sterilizing a reusable cannula. Following soaking in enzymatic soluntion the cannula should be cleaned manually with a brush and detergent or ultrasonically. Sklar cannula cleaning brushes are designed for manual cleaning and are available in several sizes in order to properly decontaminate specific size cannulas used for specific procedures. This product has a 2mm wide cleaning brush and an overall length of 12 inches. It also features a loop at the end of the handle for improved user grip and control.
Catalog No. 50-110-9713