Learn More
Instech Rat tail vein cannula, 25ga, 25cm PE/PVC tube, PinPort and injector
SDP
Supplier:  Instech C15SSRTV1438P
Encompass
The Instech C15SS-RTV1438P is a rat tail vein cannula with a sharp 25ga stainless steel introducer and a 25cm (9.8in) BTCOEX-25 PE/PVC extension tube for reliable venous access during dosing or sampling. Its integrated white 25ga PinPort connector provides a low-dead-volume closed system, and the cannula set includes a PNP3M injector and needle guard for safe, ready-to-use handling. Individually EtO sterilized, it delivers consistent performance for precision rat tail vein procedures.
Features
  • 25ga stainless steel introducer (0.25x0.51mm)
  • 25cm (9.8in) BTCOEX-25 tubing (0.4x1.3mm)
  • Integrated 25ga white PinPort connector
  • Includes PNP3M injector and needle guard
  • Approx. 40µL dead volume
  • Sharp tip for clean tail vein entry
  • Individually packaged and EtO sterilized
Catalog No. 50-890-075