Learn More
Enterprise Technology Solutions SleepSense Cannula Disposable Filter attaches to any SleepSense Pressure Cannula.
SDP
Supplier:  Enterprise Technology Solutions SLS1580925
Encompass
The SleepSense Cannula Disposable Filter attaches to any SleepSense Pressure Cannula.This low-cost hydrophobic disposable filter may be used with any type of pf cannula with the pressure sensor, eliminating the fear of patient cross-contamination and sensor contamination.
Catalog No. 50-240-7028