Learn More
Beckman Coulter Tubes and Bottles
Supplier:  Beckman Coulter 330860
Encompass_Preferred
Beckman Coulter offers a wide selection of tubes and bottles for use with their centrifuges, automated systems, and other life sciences instruments. Their polypropylene and polystyrene tubes and bottles come in a variety of sizes from 0.5mL to 250mL to meet diverse application needs. The tubes feature graduations and uniform wall thickness for improved centrifugation. The screw caps have silicone gaskets for leakproof sealing. The tubes and bottles are sterilized and RNase/DNase free for reliable sample storage. Beckman Coulter provides a convenient tube and bottle selector guide to help customers identify the optimal tubes for their application.
Catalog No. NC1168027