Learn More
Chemglass Life Sciences Chem-Thread Compression Cap with Aperture, Size #4, Accepts 2.5-4mm OD Tubing
SDP
Supplier:  Chemglass Life Sciences CG35210
Encompass_Preferred
Chem-Thread Compression Cap with Aperture, Size #4, Accepts 2.5-4mm OD Tubing. Celcon compression cap with listed hole size for Chem-Threads.
Catalog No. 50-873-757