Learn More
Trajan Scientific L-MARK Brand Cap, Screw, Blue, Pre-slit Red PTFE/White Silicone, 9mm, Case of 1000
Supplier:  Trajan Scientific LPP.00903907810
Encompass_Preferred
L-MARK Brand, Cap, Screw, Polypropylene, Blue, Pre-Slit Red PTFE/White Silicone, 9 mm, Press fit, Case of 1000. Made in the USA.
Catalog No. 50-125-0934