Learn More
Microsolv Technology Corporation Cap Screw Green Sil/Ptfe
Supplier:  Microsolv Technology Corporation 9502SE15G
Encompass_Preferred
Caps, Screw Top, with White Silicone Rubber / Natural PTFE Septa, fitted in knurled, polypropylene, Green Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 100/PK.
Catalog No. 50-014-52161