Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE Septa, fitted in smooth sided, Robo Style, soft polypropylene, Yellow Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK.
Supplier:  Microsolv Technology Corporation 9502S05Y
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE Septa, fitted in smooth sided, Robo Style, soft polypropylene, Yellow Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 50-227-1320