Learn More
Microsolv Technology Corporation Cap Screw Top Sil/Tef Lc Clean
Supplier:  Microsolv Technology Corporation 9502S07LCC
Encompass_Preferred
Caps, Screw Top, with Clear Silicone Rubber / PTFE, ultra-pure, Pre-Slit LC-MS compatible LC Clean Septa, bonded in knurled, polypropylene, Grey Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52085