Learn More
Medchemexpress LLC Capecitabine | 154361-50-9 | 99.99% | C15H22FN3O6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB001610MM/1ML
Encompass_Preferred
Capecitabine is an oral proagent that converts to its active metabolite, 5-FU, via thymidine phosphorylase. It is an anti-cancer chemotherapy drug, classified as an antimetabolite. Capecitabine is converted into 5′-deoxy-5-fluorocytidine (5′DFCR), 5′-deoxy-5-fluorouridine (5′DFUR), and 5-FU by carboxylesterases (CES1 and 2), cytidine deaminase (CDD), and thymidine phosphorylase (TP) in both the liver and tumor. Capecitabine induces a significant cytotoxic effect in vitro only at high concentrations.
  • Intended for research use only
  • Converted to active metabolite, 5-FU, in the liver and tumor
  • Classified as an antimetabolite
  • Induces cytotoxic effect in vitro at high concentrations
Catalog No. 50-004-32161