Learn More
Medchemexpress LLC Capecitabine-2',3'-cyclic carbonate | 921769-65-5 | MFCD23160273 | 98.9% | 385.34 g·mol⁻¹ | C16H20FN3O7 | 50 MG

Supplier: Medchemexpress LLC HYW61495750MG
Capecitabine-2',3'-cyclic carbonate is a synthetic 5'-deoxy-5-fluorocytidine derivative supplied for laboratory research use only. It is provided as a powdered research reagent with supporting documentation for safe handling and quality verification.
- Purity (HPLC): 98.9%.
- Cas number: 921769-65-5.
- Molecular weight: 385.34 g·mol⁻¹.
- Molecular formula: C16H20FN3O7.
- Available sizes: 5 mg-200 mg (multiple vial sizes).
- Provided with certificate of analysis and safety data sheet.
- Intended for preclinical and in vitro anticancer research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.