Learn More
Selleck Chemical LLC Capecitabine S1156-1g
SDP
Supplier:  Selleck Chemical LLC S11561G
Encompass_Preferred
Capecitabine is a tumor-selective fluoropyrimidine carbamate which achieves higher intratumoral 5-FU level with lower toxicity than 5-FU Capecitabine treatment of HCT-15 cells causes condensation of DNA and induces apoptosis
Catalog No. 50-310-8322