Learn More
Selleck Chemical LLC Capecitabine 50mg 154361-50-9 RO 09-1978
SDP
Supplier:  Selleck Chemical LLC S115650MG
Encompass_Preferred
Capecitabine (RO 09-1978) is a tumor-selective fluoropyrimidine carbamate, which achieves higher intratumoral 5-FU level with lower toxicity than 5-FU. Capecitabine treatment of HCT-15 cells causes condensation of DNA and induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0959