Learn More
Medchemexpress LLC Capecitabine (standard) | 154361-50-9 | 99.99% | C15H22FN3O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0016R50MG
Encompass_Preferred
Capecitabine (Standard) is the analytical standard of Capecitabine. This product is intended for research and analytical applications. Capecitabine is an oral proagent that is converted to its active metabolite, 5-FU, by thymidine phosphorylase.
  • Analytical standard for capecitabine
  • Intended for research and analytical applications
  • Oral proagent converted to active metabolite, 5-FU, by thymidine phosphorylase
  • Commonly used in qualitative, quantitative, and methodological research experiments in hplc, gc, and ms
Catalog No. 50-004-32047